2-Bromo-5-nitropyridine, 98%, Thermo Scientific Chemicals
2-Bromo-5-nitropyridine, 98%, Thermo Scientific Chemicals
2-Bromo-5-nitropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-nitropyridine, 98%, Thermo Scientific Chemicals

CAS: 4487-59-6 | Molecular Formula: C5H3BrN2O2 | Molecular Weight: 203.00 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3576700505 gGlass bottle
Catalog number 357670050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4487-59-6
IUPAC Name2-bromo-5-nitropyridine
Molecular FormulaC5H3BrN2O2
InChI KeyHUUFTVUBFFESEN-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point137°C to 141°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only