2,5-Dichlorobenzaldehyde, 98%
2,5-Dichlorobenzaldehyde, 98%
2,5-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,5-Dichlorobenzaldehyde, 98%

CAS: 6361-23-5 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 359940050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6361-23-5
IUPAC Name2,5-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyBUXHYMZMVMNDMG-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point54°C to 58°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only