Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron - FAQs
Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron - FAQs
View additional product information for Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron - FAQs (360110500, 360112500)
1 product FAQs found
What is the pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 μm (Cat. No. 360110500, 360112500) in Angstroms?
The pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 µm (Cat. No. 360110500, 360112500) is 60 Angstroms.