Thermo Scientific Chemicals

Triphosphopyridine nucleotide, disodium salt, 97+%, Thermo Scientific Chemicals

Catalog number: 360565000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

Triphosphopyridine nucleotide, disodium salt, 97+%, Thermo Scientific Chemicals

Catalog number: 360565000
500 mg, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 360565000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
24292-60-2
IUPAC Name
disodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-(hydrogen phosphonatooxy)-3-hydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular Formula
C21H26N7Na2O17P3
InChI Key
WSDDJLMGYRLUKR-MWLGWILCNA-L
SMILES
[Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Appearance (Color)
Yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Assay
>=97.0 % (enzymatic, on dry subst.)
Water
=<6.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text