Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2.5 kg
12 kg
100 lb.
50 kg
350 lb.
Katalognummer 3627-65
Preis (EUR)
-
Menge:
50 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Specifications
Siedepunkt1413°C (1013 hPa)
CAS7647-14-5
Chemischer Name oder MaterialSodium chloride
FarbeWeiß
Schmelzpunkt801°C
VerpackungDrum
SynonymHalite, Rock salt, Saline, Salt, Sea salt
GüteMulti-Kompendium
Molekulargewicht (g/mol)58.44
Menge50 kg
Unit SizeEach