Trisodium citrate dihydrate, 99.0-100.5% (by anhydrous basis), granular, FCC, USP, GMP, J.T.Baker™
Avantor™

Trisodium citrate dihydrate, 99.0-100.5% (by anhydrous basis), granular, FCC, USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2.5 kg
12 kg
50 kg
100 kg
Katalognummer 3649-01
Preis (EUR)
-
Menge:
500 g
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P Specifications.
Specifications
CAS4-3-6132
Chemischer Name oder MaterialTrisodium citrate
FarbeWeiß
Schmelzpunkt150°C
SummenformelNa3C6H5O7·2H2O
VerpackungAmber glass bottle, wide mouth, tamper-evident cap
SynonymSodium citrate dihydrate, Citric acid trisodium salt dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Citric acid tridosium salt dihydrate
GüteFCC
Molekulargewicht (g/mol)294.1
Reinheit (%)99.0-100.5%
Menge500 g
Unit SizeEach