2-Methylpyrrolidine, 98%
2-Methylpyrrolidine, 98%
2-Methylpyrrolidine, 98%
Thermo Scientific Chemicals

2-Methylpyrrolidine, 98%

CAS: 765-38-8 | C5H11N | 85.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
36539010010 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 365390100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS765-38-8
IUPAC Name(2S)-2-methylpyrrolidin-1-ium
Molecular FormulaC5H12N
InChI KeyRGHPCLZJAFCTIK-YFKPBYRVSA-O
SMILESC[C@H]1CCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4350 to 1.4370 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only