Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A
Thermo Scientific Chemicals

Aluminum oxide, for chromatography, acidic, Brockmann I, 40-300 μm, 60A

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 365750010
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sodium oxide (Na2O)=<0.4 %
Specific surface area185 to 225 m²/g
pH4.0 to 5.0 (10% aqueous suspension)
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
RUO – Research Use Only