N,N,N'-Trimethylethylenediamine, 97%
N,N,N'-Trimethylethylenediamine, 97%
N,N,N'-Trimethylethylenediamine, 97%
Thermo Scientific Chemicals

N,N,N'-Trimethylethylenediamine, 97%

CAS: 142-25-6 | C5H14N2 | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 366910050
Price (USD)
65.65
Online exclusive
73.30
Save 7.65
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
65.65
Online exclusive
73.30
Save 7.65
Each
Chemical Identifiers
CAS142-25-6
IUPAC Name[2-(dimethylazaniumyl)ethyl](methyl)azanium
Molecular FormulaC5H16N2
InChI KeyHVOYZOQVDYHUPF-UHFFFAOYSA-P
SMILESC[NH2+]CC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4180 to 1.4210 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only