Iodine monochloride, 1M solution in dichloromethane
Iodine monochloride, 1M solution in dichloromethane
Iodine monochloride, 1M solution in dichloromethane
Thermo Scientific Chemicals

Iodine monochloride, 1M solution in dichloromethane

CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
Glass bottle
Catalog number 370741000
Price (USD)
69.65
Online exclusive
77.20
Save 7.55
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
69.65
Online exclusive
77.20
Save 7.55
Each
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Appearance (Form)Solution
Titration with Na2S2O30.95 to 1.05 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only