Ethyl benzoylacetate, 90%
Ethyl benzoylacetate, 90%
Ethyl benzoylacetate, 90%
Thermo Scientific Chemicals

Ethyl benzoylacetate, 90%

CAS: 94-02-0 | C11H12O3 | 192.21 g/mol
数量:
50 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 370851000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS94-02-0
IUPAC Nameethyl 3-oxo-3-phenylpropanoate
Molecular FormulaC11H12O3
InChI KeyGKKZMYDNDDMXSE-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1=CC=CC=C1
さらに表示
Refractive index1.5240 to 1.5330 (20°C, 589 nm)
HPLC>=88 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to red to brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only