Bromodiphenylmethane, 90%
Bromodiphenylmethane, 90%
Bromodiphenylmethane, 90%
Thermo Scientific Chemicals

Bromodiphenylmethane, 90%

CAS: 776-74-9 | C13H11Br | 247.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 373461000
Price (EUR)
70,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
70,30
Each
Chemical Identifiers
CAS776-74-9
IUPAC Name[bromo(phenyl)methyl]benzene
Molecular FormulaC13H11Br
InChI KeyOQROAIRCEOBYJA-UHFFFAOYSA-N
SMILESBrC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline mass or solid
Appearance (Color)Orange to brown
GC>=88 %
Infrared spectrumConforms
Melting point33°C to 42°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only