Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 1 Antibodies

          Invitrogen

          Claudin 1 Monoclonal Antibody (2H10D10)

          Advanced Verification
          66 Published Figures
          128 References
          View all (32) Claudin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 1 Monoclonal Antibody (2H10D10)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (66)
          • Advanced Verification (2)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 74

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 1 Antibody (37-4900) in ICC/IF

          Immunofluorescent analysis of Claudin-1 was done on 90% confluent log phase CaCo-2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes; permeabilized with 0.25% Triton™ X-100 for 10 minutes followed by blocking with 5% BSA for 1 hour at room temperature. The cells were incubated with Claudin-1 Mouse Monoclonal Antibody (Product # 37-4900) at 1 µg/mL in 1% BSA and incubated for 3 hours at roo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (IHC)
          Claudin 1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody in Western Blot (WB)
          Claudin 1 Antibody
          Claudin 1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Claudin 1 Monoclonal Antibody (2H10D10)

          Product Details

          37-4900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 61 publications 61 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 8 publications 8 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          5-10 µg/mL
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1-5 µg/mL
          View 24 publications 24 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -

          Immunoprecipitation (IP)

          10 µg/test
          -

          Miscellaneous PubMed (Misc)

          -
          View 16 publications 16 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Chicken, Dog, Human, Mouse, Pig, Rabbit, Rat, Rhesus monkey

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2H10D10

          Immunogen

          Synthetic peptide derived from the C-terminal region of the human and mouse claudin-1 proteins
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533323

          Product Specific Information

          Reactivity has been confirmed with human Caco-2 and canine MDCK cell lysates, mouse kidney and intestinal lysates, frozen mouse intestine (jejunum) tissue, and rat kidney homogenates.

          This antibody is specific for the C-terminal region of the claudin-1 protein. On western blots, it identifies the target band at ~22 kDa.

          Target Information

          Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CLDN1; senescence-associated epithelial membrane protein 1; tight junction transmembrane protein; unnamed protein product

          View more View less

          Gene Aliases: CLD1; ILVASC; SEMP1

          View more View less

          UniProt ID: (Human) O95832, (Mouse) O88551, (Rat) P56745

          View more View less

          Entrez Gene ID: (Human) 9076, (Dog) 608207, (Mouse) 12737, (Rat) 65129

          View more View less

          Function(s)
          virus receptor activity structural molecule activity protein binding identical protein binding tight junction
          Process(es)
          establishment of blood-nerve barrier response to toxic substance calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules positive regulation of cell migration response to lipopolysaccharide cell junction maintenance maintenance of blood-brain barrier cell-cell junction organization response to ethanol viral entry into host cell protein oligomerization positive regulation of epithelial cell proliferation involved in wound healing establishment of skin barrier xenobiotic transport across blood-nerve barrier response to interleukin-18 bicellular tight junction assembly cellular response to lead ion cellular response to interferon-gamma cellular response to tumor necrosis factor response to dexamethasone cellular response to transforming growth factor beta stimulus positive regulation of wound healing establishment of endothelial intestinal barrier liver regeneration positive regulation of bicellular tight junction assembly cellular response to butyrate
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch