Thermo Scientific Chemicals

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides), Thermo Scientific Chemicals

Catalog number: 378662500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides), Thermo Scientific Chemicals

Catalog number: 378662500
250 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
13454-94-9
IUPAC Name
dicerium(3+) trisulfate
Molecular Formula
Ce2O12S3
InChI Key
OZECDDHOAMNMQI-UHFFFAOYSA-H
SMILES
[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
White to off-white
Appearance (Form)
Powder or crystals
Titration Complexometric
46.0 to 50.0 % (Ce)
Total conc. of imp.as REO
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text