Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Thermo Scientific Chemicals

Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized

CAS: 123-41-1 | C5H15NO2 | 121.18 g/mol
数量:
500 mL
2.5 L
10 L
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 386720100
価格(JPY)
-
数量:
10 L
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123-41-1
IUPAC Name(2-hydroxyethyl)trimethylazanium hydroxide
Molecular FormulaC5H15NO2
InChI KeyKIZQNNOULOCVDM-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)CCO
さらに表示
Appearance (Form)Viscous solution
Stabilizerabout 0.2 % ethylenediamine
Appearance (Color)Colorless to tan
Titration with HClO444.0 to 48.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only