Calcium perchlorate tetrahydrate, pure
Calcium perchlorate tetrahydrate, pure
Calcium perchlorate tetrahydrate, pure
Thermo Scientific Chemicals

Calcium perchlorate tetrahydrate, pure

CAS: 15627-86-8 | CaCl2O8·4H2O | 311.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
390631000100 gGlass bottle
Catalog number 390631000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Chemical Identifiers
CAS15627-86-8
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric12.2 to 13.5 % (Calcium)
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only