N,N-Dimethylacetamide, 99.8+%, for HPLC
N,N-Dimethylacetamide, 99.8+%, for HPLC
N,N-Dimethylacetamide, 99.8+%, for HPLC
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99.8+%, for HPLC

CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 390670010
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
148.65
Online exclusive
165.00
Save 16.35
Each
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.80 %
Appearance (Color)Clear colorless
Water=<0.05 %
Acidity=<0.0005 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only