6-Bromo-2-picoline, 97%
6-Bromo-2-picoline, 97%
6-Bromo-2-picoline, 97%
Thermo Scientific Chemicals

6-Bromo-2-picoline, 97%

CAS: 5315-25-3 | C6H6BrN | 172.03 g/mol
数量:
1 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 391050100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5315-25-3
IUPAC Name2-bromo-6-methylpyridine
Molecular FormulaC6H6BrN
InChI KeySOHDPICLICFSOP-UHFFFAOYSA-N
SMILESCC1=CC=CC(Br)=N1
さらに表示
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5620 to 1.5640 (20°C, 589 nm)
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only