Titanium(IV) bromide, 98%
Titanium(IV) bromide, 98%
Titanium(IV) bromide, 98%
Thermo Scientific Chemicals

Titanium(IV) bromide, 98%

CAS: 7789-68-6 | TiBr4 | 367.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 391240050
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.90
Save 8.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
54.90
Save 8.25
Each
Chemical Identifiers
CAS7789-68-6
IUPAC Nametitanium(4+) tetrabromide
Molecular FormulaBr4Ti
InChI KeyUBZYKBZMAMTNKW-UHFFFAOYSA-J
SMILES[Ti+4].[Br-].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Titration Argentometric97.5 to 102.5 %
Appearance (Form)Powder and/or lumps or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only