Chloro(triphenylphosphine)gold(I), 99%
Chloro(triphenylphosphine)gold(I), 99%
Chloro(triphenylphosphine)gold(I), 99%
Thermo Scientific Chemicals

Chloro(triphenylphosphine)gold(I), 99%

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol
数量:
250 mg
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 391612500
価格(JPY)
-
数量:
250 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14243-64-2
IUPAC Namechlorogold; triphenylphosphane
Molecular FormulaC18H15AuClP
InChI KeyIFPWCRBNZXUWGC-UHFFFAOYSA-M
SMILESCl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Assay>=98.5 % (ex Au)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only