Tetramethylammonium triacetoxyborohydride, 90+%
Tetramethylammonium triacetoxyborohydride, 90+%
Tetramethylammonium triacetoxyborohydride, 90+%
Thermo Scientific Chemicals

Tetramethylammonium triacetoxyborohydride, 90+%

CAS: 109704-53-2 | C10H22BNO6 | 263.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 392850100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS109704-53-2
IUPAC Namebis(acetyloxy)boranuidyl acetate; tetramethylazanium
Molecular FormulaC10H22BNO6
InChI KeyLCFZZOGKVOTFPU-UHFFFAOYSA-N
SMILESC[N+](C)(C)C.CC(=O)O[BH-](OC(C)=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder
Titration with HClO4>=90.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only