3-Furaldehyde, 98%, stabilized
3-Furaldehyde, 98%, stabilized
3-Furaldehyde, 98%, stabilized
Thermo Scientific Chemicals

3-Furaldehyde, 98%, stabilized

CAS: 498-60-2 | C5H4O2 | 96.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 393260250
Price (USD)
290.65
Special offer
Online exclusive
Ends: 28-Dec-2026
342.00
Save 51.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
290.65
Special offer
Online exclusive
Ends: 28-Dec-2026
342.00
Save 51.35
Each
Chemical Identifiers
CAS498-60-2
IUPAC Namefuran-3-carbaldehyde
Molecular FormulaC5H4O2
InChI KeyAZVSIHIBYRHSLB-UHFFFAOYSA-N
SMILESO=CC1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
GC>=97.5 %
Stabilizer0.05 % BHT
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only