Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
Thermo Scientific Chemicals

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid

CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 395620010
Price (USD)
385.65
Special offer
Online exclusive
Ends: 28-Dec-2026
453.00
Save 67.35
Each
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
385.65
Special offer
Online exclusive
Ends: 28-Dec-2026
453.00
Save 67.35
Each
Chemical Identifiers
CAS7705-07-9
IUPAC Nametitanium(3+) trichloride
Molecular FormulaCl3Ti
InChI KeyYONPGGFAJWQGJC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ti+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark purple
Appearance (Form)Solution
Titration Complexometric18 to 23 %
Titration with NaOH1.5 to 2.5 N HCl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only