2,6-Dichloronicotinic acid, 98+%
2,6-Dichloronicotinic acid, 98+%
2,6-Dichloronicotinic acid, 98+%
Thermo Scientific Chemicals

2,6-Dichloronicotinic acid, 98+%

CAS: 38496-18-3 | C6H3Cl2NO2 | 192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 396610050
Price (USD)
92.70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
92.70
Each
Chemical Identifiers
CAS38496-18-3
IUPAC Name2,6-dichloropyridine-3-carboxylic acid
Molecular FormulaC6H3Cl2NO2
InChI KeyAJPKQSSFYHPYMH-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Cl)N=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Crystalline powder
Titration with NaOH>=98.0 %
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only