Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol
製品番号(カタログ番号)数量パッケージ
397611000100 mLAcroSeal™ Glass Bottle
製品番号(カタログ番号) 397611000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS23719-80-4
IUPAC Namebromo(cyclopropyl)magnesium
Molecular FormulaC3H5BrMg
InChI KeyQUPLLJGAJVXXBW-UHFFFAOYSA-M
SMILESBr[Mg]C1CC1
さらに表示
Titration Grignard0.4 to 0.6 M
Appearance (Color)Colorless to light yellow to dark yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only