2-Bromomalonaldehyde, 97+%
2-Bromomalonaldehyde, 97+%
2-Bromomalonaldehyde, 97+%
Thermo Scientific Chemicals

2-Bromomalonaldehyde, 97+%

CAS: 2065-75-0 | C3H3BrO2 | 150.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 397900050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2065-75-0
IUPAC Name2-bromopropanedial
Molecular FormulaC3H3BrO2
InChI KeySURMYNZXHKLDFO-UHFFFAOYSA-N
SMILESBrC(C=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only