Racks for the CryoMed™ Controlled-Rate Freezer
Actual product may vary
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Thermo Scientific™

Racks for the CryoMed™ Controlled-Rate Freezer

More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsType
4000707161 4 mL/5 mL AmpulesVial Rack (Large)
185089Adjustable, 10-positionCanister Rack
400029440 0.25/0.5 mL StrawsStraw Rack
4000303162 Canes, 5 to 6 Ampules/CaneCane Freezing Rack
400031048 Skin Packets (2 level)Skin Rack
400031172 Packets (3 level)Skin Rack
400031224 Packets (1 level)Skin Rack
400034010 Canisters (for cord blood, Model 4000610)Canister Rack
400070077 CanesCane Freezing Rack
400070176 1.2 mL/2 mL AmpulesVial Rack (Small)
40007025 x 1.2/2 mL Cryo Racks (Small)Vial Rack Holder
4000703161 1.2 mL/2 mL AmpulesVial Rack (Large)
40007045 x 1.2/2 mL Cryo Racks (Large)Vial Rack Holder
400070576 4 mL/5 mL AmpulesVial Rack (Small)
40007063 x 4/5 mL Cryo Racks (Small)Vial Rack Holder
40007083 x 4/5 mL Cryo Racks (Large)Vial Rack Holder
Catalog number 4000707
Price (BRL)
-
Holds:
161 4 mL/5 mL Ampules
Type:
Vial Rack (Large)

More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.

  • Warranty varies by country, please contact us for details
Specifications
Description161 4 mL/5 mL Ampules
For Use With (Equipment)CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458
Holds161 4 mL/5 mL Ampules
ColorSilver
TypeVial Rack (Large)
Unit SizeEach