8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech.
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech.
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech.
Acros Organics

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech.

CAS: 5460-09-3 | C10H8NNaO7S2·H2O | 359.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 400840050
also known as 40084
HSN 29222190
Price (INR)
3,740.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,740.00
Each
Chemical Identifiers
CAS343321-59-5
IUPAC Namesodium 4-amino-5-hydroxy-7-sulfonaphthalene-2-sulfonate hydrate
Molecular FormulaC10H10NNaO8S2
InChI KeyYYIGOAMEITZADH-UHFFFAOYSA-M
SMILESO.[Na+].NC1=CC(=CC2=CC(=CC(O)=C12)S(O)(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to brown
Appearance (Form)Powder or crystals and/or lumps
Infrared spectrumConforms
HPLC>=75 %
Loss on drying=<6 % (1 g, 150°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only