Product Image
Applied Biosystems™

Phosphalink™, for 3' and 5' Labeling, 70 mg

Have Questions?
Catalog NumberQuantity
40171770 mg
Catalog number 401717
Price (USD)
402.00
Each
Quantity:
70 mg
Price (USD)
402.00
Each
Phosphalink™ is used for 3’ or 5’ attachment of phosphorous group during oligonucleotide synthesis.

We offer several phosphoramidites that impart the means to add a phosphate during oligonucleotide synthesis.

• Specialty phosphoramidites allow online addition of linkers during synthesis, saving time in production of labeled probes and primers.
• Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis.
• We offer a range of specialty linkers and phosphoramites to give you the flexibility demanded in oligonucleotide synthesis.

Phosphate linker:
Phosphalink™ phosphoramidite can incorporate a phosphate group at either the 5' or the 3' end. A 5'-end phosphate permits ligation of an oligonucleotide to other DNA.

Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPhosphalink™ Reagent
Column TypeOligonucleotide Synthesis Column
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormPowder
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product LinePhosphalink
Product TypeOligonucleotide Synthesis Reagent
Quantity70 mg
Reagent TypeSolvent or General Reagent
Sample TypeDNA
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in a cool and dry place.