Bis(2-ethylhexyl) terephthalate, 97%
Bis(2-ethylhexyl) terephthalate, 97%
Bis(2-ethylhexyl) terephthalate, 97%
Thermo Scientific Chemicals

Bis(2-ethylhexyl) terephthalate, 97%

CAS: 6422-86-2 | C24H38O4 | 390.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 402490010
Price (USD)
61.65
Online exclusive
68.80
Save 7.15
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.65
Online exclusive
68.80
Save 7.15
Each
Chemical Identifiers
CAS6422-86-2
IUPAC Name1,4-bis(2-ethylhexyl) benzene-1,4-dicarboxylate
Molecular FormulaC24H38O4
InChI KeyRWPICVVBGZBXNA-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to slightly yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4870 to 1.4920 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only