Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Activated charcoal, NORIT™ CN 1, decolorizing
Thermo Scientific Chemicals

Activated charcoal, NORIT™ CN 1, decolorizing

CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 404045000
Price (USD)
65.60
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
65.60
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Powder
pH5.5 to 8
Phosphate (PO4)=<3.5 % (by acid extraction)
Calcium (Ca)=<200 ppm (by acid extraction)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only