Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent

CAS: 19004-19-4 | 2·5 H2O | 232.6 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 405850025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Calcium (Ca)=<50 ppm
Lead (Pb)=<10 ppm
Potassium (K)=<50 ppm
Appearance (Form)Crystalline powder
Insoluble matter=<0.01 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only