Dibutyl phosphate, 97%
Dibutyl phosphate, 97%
Dibutyl phosphate, 97%
Thermo Scientific Chemicals

Dibutyl phosphate, 97%

CAS: 107-66-4 | C8H19O4P | 210.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 406642500
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.90
Save 8.25
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.90
Save 8.25
Each
Chemical Identifiers
CAS107-66-4
IUPAC Namedibutoxyphosphinic acid
Molecular FormulaC8H19O4P
InChI KeyJYFHYPJRHGVZDY-UHFFFAOYSA-N
SMILESCCCCOP(O)(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=96.0 %
Water=<0.3 %
Appearance (Form)Clear liquid
Color scale=<100 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only