Dibutyl sebacate, 93%
Dibutyl sebacate, 93%
Dibutyl sebacate, 93%
Thermo Scientific Chemicals

Dibutyl sebacate, 93%

CAS: 109-43-3 | C18H34O4 | 314.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 406652500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-43-3
IUPAC Name1,10-dibutyl decanedioate
Molecular FormulaC18H34O4
InChI KeyPYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCCCCCCCC(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<40 APHA
Infrared spectrumConforms
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only