Thermo Scientific Chemicals

4-Ethoxyphenol, 99%, Thermo Scientific Chemicals

Catalog number: 409670250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Ethoxyphenol, 99%, Thermo Scientific Chemicals

Catalog number: 409670250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
622-62-8
IUPAC Name
4-ethoxyphenol
Molecular Formula
C8H10O2
InChI Key
LKVFCSWBKOVHAH-UHFFFAOYSA-N
SMILES
CCOC1=CC=C(O)C=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks
Melting point
64°C to 67°C
Appearance (Color)
Beige
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text