Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Thermo Scientific Chemicals

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%

CAS: 67-42-5 | C14H24N2O10 | 380.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 409911000
Price (EUR)
301,65
Online exclusive
335,00
Save 33,35 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
301,65
Online exclusive
335,00
Save 33,35 (10%)
Each
Chemical Identifiers
CAS67-42-5
IUPAC Name3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular FormulaC14H24N2O10
InChI KeyDEFVIWRASFVYLL-UHFFFAOYSA-N
SMILESOC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=97.5 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point240°C to 244°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only