Methylene di-p-phenyl diisocyanate, 98%, flakes
Methylene di-p-phenyl diisocyanate, 98%, flakes
Methylene di-p-phenyl diisocyanate, 98%, flakes
Thermo Scientific Chemicals

Methylene di-p-phenyl diisocyanate, 98%, flakes

CAS: 101-68-8 | C15H10N2O2 | 250.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 414285000
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Chemical Identifiers
CAS101-68-8
IUPAC Name1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
Molecular FormulaC15H10N2O2
InChI KeyUPMLOUAZCHDJJD-UHFFFAOYSA-N
SMILESO=C=NC1=CC=C(CC2=CC=C(C=C2)N=C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Flakes
Melting point39°C to 44°C
Titration>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only