Methyl Red, sodium salt, ACS reagent
Methyl Red, sodium salt, ACS reagent
Methyl Red, sodium salt, ACS reagent
Thermo Scientific Chemicals

Methyl Red, sodium salt, ACS reagent

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 414840100
Price (USD)
27.65
Online exclusive
30.20
Save 2.55
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
27.65
Online exclusive
30.20
Save 2.55
Each
Chemical Identifiers
CAS845-10-3
IUPAC Namesodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC15H14N3NaO2
InChI KeyGNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Clarity of solution(Aqueous) Passes test
Appearance (Color)Orange to red to brown
Clarity of alcohol solutionPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only