Nitro Blue Tetrazolium Chloride, 90%
Nitro Blue Tetrazolium Chloride, 90%
Nitro Blue Tetrazolium Chloride, 90%
Thermo Scientific Chemicals

Nitro Blue Tetrazolium Chloride, 90%

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 415920010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS298-83-9
IUPAC Name2-{3,3'-dimethoxy-4'-[3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-[1,1'-biphenyl]-4-yl}-3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular FormulaC40H30Cl2N10O6
InChI KeyFSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=88 %
Loss on drying=<10.0 % (105°C)
Sulfated ash=<0.3 %
Infrared spectrumConforms
Appearance (Color)Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only