Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Thermo Scientific Chemicals

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol
数量:
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 419430100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10034-88-5
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
さらに表示
Appearance (Form)Adhering crystals
Titration with NaOH>=96.0 %
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<50 ppm
Appearance (Color)White
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only