Sodium methoxide, pure, titrant, (0.5 M in methanol)
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Thermo Scientific Chemicals

Sodium methoxide, pure, titrant, (0.5 M in methanol)

CAS: 124-41-4 | CH3NaO | 54.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 419605000
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
191.65
Online exclusive
213.00
Save 21.35
Each
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Strength0.45 to 0.60 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only