Sodium periodate, 99.8+%, ACS reagent
Sodium periodate, 99.8+%, ACS reagent
Sodium periodate, 99.8+%, ACS reagent
Thermo Scientific Chemicals

Sodium periodate, 99.8+%, ACS reagent

CAS: 7790-28-5 | INaO4 | 213.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 419610025
Price (USD)
2,210.00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
2,210.00
Each
Chemical Identifiers
CAS7790-28-5
IUPAC Namesodium periodate
Molecular FormulaINaO4
InChI KeyJQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Other halogens (as Cl)=<0.02 %
Appearance (Form)Crystalline powder
Titration Iodometric99.8 to 100.3 % (on dried basis)
Manganese (Mn)=<3 ppm
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only