2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%
2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%
2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%
Thermo Scientific Chemicals

2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%

CAS: 25265-77-4 | C12H24O3 | 216.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 422010025
Price (USD)
91.65
Online exclusive
102.00
Save 10.35
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
91.65
Online exclusive
102.00
Save 10.35
Each
Chemical Identifiers
CAS25265-77-4
IUPAC Name3-hydroxy-2,2,4-trimethylpentyl 2-methylpropanoate
Molecular FormulaC12H24O3
InChI KeyDAFHKNAQFPVRKR-UHFFFAOYNA-N
SMILESCC(C)C(O)C(C)(C)COC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4410 to 1.4430 (20°C, 589 nm)
Color scale=<10 APHA
GC>=98.0 % (sum two isomers)
Appearance (Form)Clear liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only