Ammonium hydroxide, ACS reagent, 28-30% solution in water
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Ammonium hydroxide, ACS reagent, 28-30% solution in water
Thermo Scientific Chemicals

Ammonium hydroxide, ACS reagent, 28-30% solution in water

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
10 L
Catalog number 423305000
Price (USD)
56.65
Online exclusive
63.20
Save 6.55
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
56.65
Online exclusive
63.20
Save 6.55
Each
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
AppearanceFree from suspended matter or sediment
Titration with HCl28.0 to 30.0 % (as NH3)
Chloride (Cl)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only