Ammonium phosphate, dibasic, 98+%, ACS reagent
Ammonium phosphate, dibasic, 98+%, ACS reagent
Ammonium phosphate, dibasic, 98+%, ACS reagent
Thermo Scientific Chemicals

Ammonium phosphate, dibasic, 98+%, ACS reagent

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423370025
Price (USD)
384.65
Online exclusive
427.00
Save 42.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
384.65
Online exclusive
427.00
Save 42.35
Each
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration with HCl>=98.0 %
Heavy metals (as Pb)=<0.001 %
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only