Thermo Scientific Chemicals

Ammonium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423400250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423400250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423400250
Price (USD)
Price: 53.00
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-20-2
IUPAC Name
sulfuric acid diamine
Molecular Formula
H8N2O4S
InChI Key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=O
Insoluble matter
=<0.005 %
pH
5.0 to 6.0 (5 % at 25.0°C)
Residue after ignition
=<0.005 %
Chloride (Cl)
=<5 ppm
Nitrate (NO3)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text