Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Thermo Scientific Chemicals

Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur.

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
4 L
25 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number 423685000
Price (USD)
73.65
Online exclusive
81.20
Save 7.55
Each
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (USD)
73.65
Online exclusive
81.20
Save 7.55
Each
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.5 %
Water=<0.2 % (K.F.)
Density(20/20°C) 0.901 to 0.904
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only