Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals

Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
25 L
Packaging:
View all
Fiberboard Box
Catalog number 423950200
Price (USD)
351.65
Special offer
Online exclusive
Ends: 28-Dec-2026
414.00
Save 62.35
Each
Quantity:
20 L
Packaging:
Fiberboard Box
Request bulk or custom format
Price (USD)
351.65
Special offer
Online exclusive
Ends: 28-Dec-2026
414.00
Save 62.35
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.8 %
Water=<0.1 % (K.F.)
Density(20/4°C) 0.791 to 0.793
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only