Phosphoric acid, ACS reagent, 85+% solution in water
Phosphoric acid, ACS reagent, 85+% solution in water
Phosphoric acid, ACS reagent, 85+% solution in water
Thermo Scientific Chemicals

Phosphoric acid, ACS reagent, 85+% solution in water

CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 424040025
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
200.00
Save 29.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
200.00
Save 29.35
Each
Chemical Identifiers
CAS7664-38-2
IUPAC Namephosphoric acid
Molecular FormulaH3O4P
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILESOP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.001 %
Heavy metals (as Pb)=<10 ppm
Chloride (Cl)=<3 ppm
Manganese (Mn)=<0.5 ppm
Sulfate (SO4)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only