Potassium carbonate, 99+%, ACS reagent, anhydrous
Potassium carbonate, 99+%, ACS reagent, anhydrous
Potassium carbonate, 99+%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Potassium carbonate, 99+%, ACS reagent, anhydrous

CAS: 584-08-7 | CK2O3 | 138.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 424081000
Price (USD)
42.80
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
42.80
Each
Chemical Identifiers
CAS584-08-7
IUPAC Namedipotassium carbonate
Molecular FormulaCK2O3
InChI KeyBWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Granular powder
Titration with HCl>=99.0 %
Insoluble matter=<0.01 %
Chloride (Cl)=<0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only